Learn More
Thermo Scientific™ Eutech™ pH 700 Meter
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools.
Brand: Thermo Scientific™ 01X541601
Code : A0
Additional Details : Weight : 0.10000kg
Description
Economical, user-friendly and accurate, the Eutech pH 700 is your ideal choice for routine applications in laboratories, productions plants and schools. It features a large, easy-to-read display, ready indicator that letsyou know when readings are stable and up to 5-point calibration with auto-buffer recognition. It also has a non-volatile memory holds up to 100 data points and comes witu an integral electrode holder.
Applications:
- Educational: Useful for most laboratory, ecological studies and other applications.
- Laboratory: Environmental studies, chemical labs, titrations and quality assurance testing. Use in all types of food processing.
Buffer Sets
Specifications
Eutech pH 700 Meter with 100/240 VAC Adapter | |
±0.3°C/±0.5°F | |
0.01 | |
Up to 5 | |
32.0°F to 212.0°F | |
Offset in 0.1 °increments; offset range: ±5.0 °C / ±9.0 °F | |
ORP: ±0.2 mV (±199.9 mV) / ±2 mV (beyond) | |
±2000 mV | |
ATC/MTC (0°C to 100°C) (pH only) |
±0.01 | |
-2 to 16 | |
±2000 mV | |
0°C to 100°C | |
0.1°C/°F | |
9 V DC adapter, 1.3 A | |
100/240 VAC, SMPS | |
ORP: 0.1 mV (±199.9 mV) / 1 mV (beyond) |