missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Protein Cleaning Solution for pH electrodes

Product Code. 12941678
Change view
Click to view available options
For Use With (Equipment):
pH Meter, Electrode
Unit Size:
480mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12941678 pH Meter, Electrode 480mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12941678 Supplier Thermo Scientific™ Supplier No. 01X211216
  • ONLINE EXCLUSIVE PRICE
    39.35 €
    37.00 € / 480mL
    Save 2.35 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH buffers and solutions for use with pH meters and electrodes

The measure of a liquid’s acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications.

Eutech™ offers a wide range of pH buffers, calibration, and cleaning solutions for use with pH meters and electrodes.

Features:

  • Buffers, calibration and cleaning solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

Description Protein cleaning solution for pH electrodes
For Use With (Equipment) pH Meter, Electrode
Type pH Electrode Protein Cleaning Solution
Quantity 450 mL
compliance-icons compliance-icons
Product Identifier
  • protein cleaning solution
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash skin thoroughly after handling.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid, pepsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.