missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ Salinity (NaCl) Standard Solutions

Product Code. 15601717 Shop All Thermo Scientific Products
Change view
Click to view available options
Concentration:
25 ppt
5 ppt
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration Unit Size
15601717 5 ppt Each
16445339 25 ppt Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15601717 Supplier Thermo Scientific™ Supplier No. 01X211230
  • ONLINE EXCLUSIVE PRICE
    17.40 €
    16.80 € / Pack of 1
    Save 0.60 €
    3% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers Salinity Standard Solutions for use with Salinity Meters.

Eutech™ offers a range of Salinity Standard Solutions for use with Salinity Meters. Salinity measurements are common in industries like agriculture, aquaculture, hydroponics, food, pools and spas where it is necessary to monitor the salt level constantly.

Features:

  • Buffers and calibration solutions packed in 480 ml durable plastic bottles – economical and convenient for laboratory use.

Description:

  • ECNACL5PPT: 5 ppt (parts per thousand) Nacl calibration solution
  • ECNACL25PPT: 25 ppt (parts per thousand) Nacl calibration solution
  • ECNACL45PPT: 45 ppt(parts per thousand) Nacl calibration solution
TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 ppt
Description 5 ppt NaCl Calibration Solution
Type NaCl Calibration Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.