missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 6+ Meter

Product Code. 11569164
Change view
Click to view available options
Product Type:
pH Meter with ATC Electrode
pH Meter with Carrying Kit
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11569164 pH Meter with Carrying Kit Each
11549164 pH Meter with ATC Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11569164 Supplier Thermo Scientific™ Supplier No. 01X245028
  • ONLINE EXCLUSIVE PRICE
    478.00 €
    454.00 € / Pack of 1
    Save 24.00 €
    5% Off
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

The measure of a liquid's acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. The Eutech™ pH 6+ Meter is an ideal choice for pH applications from the wide range of pH meters that Eutech™ offers.

Features:

  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
TRUSTED_SUSTAINABILITY

Specifications

Accuracy (mV) ± 1 mV
Accuracy (pH) ±0.01
Battery Type 4 AAA x 1.4 V
Calibration Points Up to 5
Description Eutech pH 6+ pH Meter with ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised Rinse Water -60 ml Each), Rubber Armour/Stand
Display Type LCD
Measurement Modes Acidity, Alkalinity, Conductivity, Temperature, pH
Range (mV) -1000 to 1000 mV
Range (pH) 0 to 14
Resolution (pH) 0.01
Temperature Range (Metric) 0°C to 100.0°C
Includes Auto Power-Off after 17 minutes; Hold & Self-diagnostic messages
Product Type pH Meter with Carrying Kit
Resolution (mV) 1 mV
Temperature Accuracy ±0.05°C
Temperature Compensation Automatic / Manual (0 to 100 °C)
Temperature Resolution 0.1 °C
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.