missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 6+ pH Meter

Product Code. 10548135
Change view
Click to view available options
Type:
Meter with 3-in-1 Combi Electrode
Meter with pH Electrode
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10548135 Meter with pH Electrode Each
15716275 Meter with pH Electrode Each
11559164 Meter with 3-in-1 Combi Electrode Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10548135 Supplier Thermo Scientific™ Supplier No. 01X245026
  • ONLINE EXCLUSIVE PRICE
    535.00 €
    346.67 € / Pack of 1
    Save 188.33 €
    35% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 6+ pH Meter ECPH602PLUSK with single Junction pH electrode and ECPH603PLUSK with 3-in-1 pH/Temperature Combi Electrode are ideal choices for routine applications in laboratories, productions plants and schools.

Features:

  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
TRUSTED_SUSTAINABILITY

Specifications

Description Eutech pH 6+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each), Rubber Armour/Stand
Includes Auto Power-Off after 17 minutes; Hold & Self-diagnostic messages
Accuracy (Temperature) ±0.05°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
Range (pH) 0 to 14
Range (mV) -1000 to 1000
Resolution (mV) 1 mV
Calibration Points Up to 5
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Temperature Compensation Automatic/Manual (0°C to 100°C)
Battery Type 4 AAA x 1.4V batteries
Type Meter with pH Electrode
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.