missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ pH 6+ pH Meter

Product Code. 15716275
Click to view available options
Type:
Meter with 3-in-1 Combi Electrode
Meter with pH Electrode
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15716275

Brand: Thermo Scientific™ 01X245026

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications.

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. Eutech™ pH 6+ pH Meter ECPH602PLUSK with single Junction pH electrode and ECPH603PLUSK with 3-in-1 pH/Temperature Combi Electrode are ideal choices for routine applications in laboratories, productions plants and schools.

Features:

  • Multi-Point Calibration (up to 5 points) with Auto-Buffer Recognition - choice of USA, NIST or pure water buffer standards with ±0.01 pH accuracy
  • Automatic Temperature Compensation (ATC) for the highest accuracy under fluctuating temperatures
  • Hold Function momentarily freezes reading for easy viewing and recording
  • Auto-Power Off saves battery power after non-use
  • Self-Diagnostic with message codes for easy troubleshooting
  • Large Custom LCD provides optimum viewing even at a distance
  • Protective Rubber Boot shields meter from accidental knocks and features a sturdy built-in stand for easy bench-top operation
  • Rugged Carrying Case (Optional) provides handy on-site meter calibration and measurement
  • pH Buffer Options: 1.68, 4.01, 7.00, 10.01, 12.45 (USA),1.68, 4.01, 6.86, 9.18, 12.45 (NIST), 4.10, 6.97 (Pb)
TRUSTED_SUSTAINABILITY

Specifications

Description Eutech pH 6+ pH Meter with Single Junction pH Electrode ECFC7252101B, ATC Electrode ECPHTEM01P & pH Carrying Kit Set (Includes 1 x pH 4.01, pH 7.00 Buffer Solutions, Storage Solution & 1 x Deionised (Rinse) Water (60 ml Each), Rubber Armour/Stand
Includes Auto Power-Off after 17 minutes; Hold & Self-diagnostic messages
Accuracy (Temperature) ±0.05°C
Accuracy (mV) ±1
Accuracy (pH) ±0.01
Range (pH) 0 to 14
Range (mV) -1000 to 1000
Resolution (mV) 1 mV
Calibration Points Up to 5
Temperature Range (Metric) 0°C to 100°C
Resolution (Temperature) 0.1°C
Display Type LCD
Temperature Compensation Automatic/Manual (0°C to 100°C)
Battery Type 4 AAA x 1.4V batteries
Type Meter with pH Electrode
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.