missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ EVA Foam Ice Pans and Buckets with Lids

Product Code. 16554241 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Capacity (Metric):
1 L
2.5 L
4 L
9 L
Color:
Blue
Green
Purple
Includes:
Bucket with Lid
Pan with Lid
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color Includes Unit Size
16554241 1 L Purple Pan with Lid Each
16534241 1 L Blue Pan with Lid Each
16544241 1 L Green Pan with Lid Each
16564241 4 L Blue Pan with Lid Each
16574241 4 L Green Pan with Lid Each
16584241 4 L Purple Pan with Lid Each
16594241 9 L Blue Pan with Lid Each
16504251 9 L Green Pan with Lid Each
16514251 9 L Purple Pan with Lid Each
16524251 2.5 L Blue Bucket with Lid Each
16534251 2.5 L Green Bucket with Lid Each
16544251 2.5 L Purple Bucket with Lid Each
16554251 4 L Blue Bucket with Lid Each
16564251 4 L Green Bucket with Lid Each
16574251 4 L Purple Bucket with Lid Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 16554241
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

Includes

removable lid.

Fisherbrand™EVA Foam Ice Pans and Buckets with Lids provide excellent insulation for any benchtop cryogenic application, without the mess!

  • Sweat-proof ethylene vinyl acetate will not leak
  • Compatible with ice, dry ice, liquid nitrogen, alcohol, and saline solutions
  • Bases and lids feature glove-friendly handles
  • Cleanup is quick and easy with molded pour spouts

Especificaciones

For Use With (Equipment) Ice, dry ice, liquid nitrogen, alcohol, and saline solutions
Includes Pan with Lid
Description Pan with Lid
Material EVA
Product Type Cryo-Safe Cold Box
Quantity 1
Color Purple
Capacity (Metric) 1 L
Autoclavable Not Autoclavable
Temperature Control No
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.