missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Evans Blue

Product Code. 16488606
Encompass_Preferred
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16488606 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16488606 Supplier MedChemExpress Supplier No. HYB1102
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Evans Blue is a potent inhibitor of L-glutamate uptake via the membrane bound excitatory amino acid transporter (EAAT).

Specifications

Product Type Biochemical Assay Reagents
CAS 314-13-6
Synonym Direct Blue 53 ∣ C.I. 23860
Solubility H2O : 33.33 mg/mL (34.69 mM; Need ultrasonic) ∣ DMSO : ≥ 25 mg/mL (26.02 mM)
Content And Storage 4°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Quantity 100 mg
Molecular Weight (g/mol) 960.81 g/mol
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.