Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint
Product Code. 11953381
Change view
Click to view available options
Standard Taper Joint Side:
NS 14/23
NS 29/32
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Side Unit Size
11953381 NS 14/23 Pack of 10
11886104 NS 29/32 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11953381 Supplier Lenz Laborglasintrumente Supplier No. 03041410
  • ONLINE EXCLUSIVE PRICE
    201.00 €
    137.60 € / Pack of 10
    Save 63.40 €
    31% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • Pear-shaped
  • With inscription label
  • Material: DURAN™
Specifications
Material Duran Glass
Standard Taper Joint Side NS 14/23
Volume (Metric) 5 mL