missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Everolimus

Product Code. 17190172
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190172 25 mg 25mg
17180172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17190172 Supplier STEMCELL Technologies Supplier No. 73124
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

mTOR pathway inhibitor; Inhibits FKBP-12

  • Everolimus is an inhibitor of the mammalian target of rapamycin (mTOR)
  • It is a hydroxyethyl ether-substituted derivative of rapamycin with improved pharmacokinetic and pharmacodynamic properties
  • It inhibits both mTORC1 and mTORC2 complexes by binding to FK506-binding protein (FKBP-12), which then binds to mTOR, leading to complex destabilization and blocked kinase function (Zeng et al.; Lebwohl et al.; Sedrani et al.; Huang & Houghton)
  • Inhibits cell proliferation, metabolism, and angiogenesis in a variety of cancers using in vitro and in vivo models (Zhu et al.; Lane et al.; O'Reilly et al.; Lebwohl et al.)
  • Acts as an immunosuppressive agent in the context of organ transplantation (Lebwohl et al.; Wullschleger et al.)

Specifications

CAS 159351-69-6
Synonym RAD001; SDZ-RAD; Xience
Quantity 25 mg
Molecular Formula C53H83NO14
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.