Learn More
Everolimus, Thermo Scientific™
A rapamycin analog
248.00 € - 412.00 €
Chemical Identifiers
| CAS | 159351-69-6 |
|---|---|
| Molecular Formula | C53H83NO14 |
| Molecular Weight (g/mol) | 958.24 |
| MDL Number | MFCD07785165 |
| InChI Key | HKVAMNSJSFKALM-PWMUVMERSA-N |
| PubChem CID | 129317877 |
| SMILES | CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15482959
|
Thermo Scientific Alfa Aesar
J60139.MA |
10mg |
248.00 €
10mg |
In Stock
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
|
15492959
|
Thermo Scientific Alfa Aesar
J60139.MB |
25mg |
412.00 €
25mg |
Estimated Shipment: 23-09-2026
Log in to see stock. |
|||||
|
You must Sign In and have a business account to purchase this item. Restricted Item |
|||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
| 159351-69-6 | |
| 958.24 | |
| HKVAMNSJSFKALM-PWMUVMERSA-N | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC |
| C53H83NO14 | |
| MFCD07785165 | |
| 129317877 |
Specifications
| 159351-69-6 | |
| MFCD07785165 | |
| Soluble in dimethysulfoxide,ethanol and chloroform; Slightly soluble in water. | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC | |
| 129317877 | |
| Everolimus |
| C53H83NO14 | |
| 14,3907 | |
| HKVAMNSJSFKALM-PWMUVMERSA-N | |
| 958.24 | |
| 958.22 |
Safety and Handling
GHS H Statement
H372
Causes damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P314-P501c
H372
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.