accessibility menu, dialog, popup

UserName

H3CO(R)(S)(R)OOH(R)CH3(S)O(R)H3CCH3(R)OH(R)OH3CO(R)CH3(S)CH3CH3(S)(S)(R)H3C(R)HOOOON(S)OOOH3C(Z)(Z)(Z)(Z)

CAS RN 159351-69-6

IUPAC Name: (1R,9S,12S,15R,16Z,18R,19R,21R,23S,24E,26Z,28E,30S,32S,35R)-1,18-dihydroxy-12-[(2R)-1-[(1S,3R,4R)-4-(2-hydroxyethoxy)-3-methoxycyclohexyl]propan-2-yl]-19,30-dimethoxy-15,17,21,23,29,35-hexamethyl-11,36-dioxa-4-azatricyclo[30.3.1.0⁴,⁹]hexatriaconta-16,24,26,28-tetraene-2,3,10,14,20-pentone
Synonyms: everolimus sdz rad RAD everolimus (substance) certican sdz-rad everolimus (product)
Molecular Weight (g/mol): 958.24
Molecular Formula: C53H83NO14
InChi Key: HKVAMNSJSFKALM-CNPAPGRKSA-N
SMILES: CO[C@@H]1C[C@H](C[C@@H](C)[C@@H]2CC(=O)[C@H](C)\C=C(C)/[C@@H](O)[C@@H](OC)C(=O)[C@H](C)C[C@H](C)\C=C\C=C/C=C(C)/[C@H](C[C@@H]3CC[C@@H](C)[C@@](O)(O3)C(=O)C(=O)N3CCCC[C@H]3C(=O)O2)OC)CC[C@H]1OCCO
Molecular Weight (g/mol) 
  • (7)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
Color 
  • (4)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (7)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (6)
  • (4)
  • (4)
  • (1)
  • (1)