Learn More
Emma Safety Footwear Everon S2 Shoe
Product Code. 17592556
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17592556 47 1 pair
17582546 36 1 pair
17592546 37 1 pair
17502556 38 1 pair
17512556 39 1 pair
17522556 40 1 pair
17532556 41 1 pair
17542556 42 1 pair
17552556 43 1 pair
17562556 44 1 pair
17572556 45 1 pair
17582556 46 1 pair
17502566 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 17592556 Supplier Emma Safety Footwear Supplier No. MM74854747
  • ONLINE EXCLUSIVE PRICE
    144.90 €
    110.36 € / 1 pairs
    Save 34.54 €
    23% Off
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Anatomically designed for a superior fit, the Everon is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
  • S2-Safety shoe for wet working conditions
  • Metal Free
  • The energy returning Everon. Anatomically designed for a superior fit, the Everon is a shoe that's been designed for long days in and out of truck cabs, building sites, warehouses - wherever your work takes you.
  • The shoe's tongue has been ingeniously designed to prevent moisture and dirt entering.
  • Comfortable, supportive with energy returning soles, the Everon's a mid-boot version of the Evoke.
  • Industries:
    • Logistics & transport
Specifications
Closure Type Laces
Color Black, Gray
Liner Material Hydro-Tec Antibacterial
Material Nubuck
Size 47
Disposable Reusable
Certifications/Compliance (Europe) ISO 20345:2011
Fit Standard Width
EN ISO 20345 S2