Learn More
Thermo Scientific™ Evodiamine
Brand:  Thermo Scientific™ J62771.MC
Quantity 100 mg
 View more versions of this product
Product Code. 15455069
  • 70.30 € / 100mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mg
-30°C to -10°C
A vanilloid receptor agonist that induces apoptosis in leukemia U937 cells
C19H17N3O
Off-white to pale yellow
Solid
518-17-2
303.40
Product Suggestions
Product Suggestions
Videos
SDS Documents