Learn More
Univet™ Evolution Disposable Sights and Frames Package
Product Code. 17981712
Change view
Click to view available options
Lens Material:
Polyester
Unit Size:
Each
Product Code. Lens Material Unit Size
17981712 Polyester Each
1 options
Product Code. 17981712 Supplier Univet™ Supplier No. MDU7003
  • ONLINE EXCLUSIVE PRICE
    25.99 €
    21.93 € / Pack of 1
    Save 4.06 €
    15% Off
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Evolution model is equipped with very light frames in different colors and with foldable temples to make it possible to put the product in the pocket after removing the disposable visor with anti-fog coating.
  • Contains 10 kits for a total of 50 sights and 10 frames
  • Each kit consists of 5 interchangeable disposable sights and 1 colored frame
  • Foldable
  • Anti-fog and disposable lens
  • Interchangeable visor
  • Ultra-light weight
  • 5 colorful frames
Specifications
Shape Oval
Lens Coating Scratch-resistant
Lens Tint None
Product Type Eyeshield
No. per Pack 10 Kits/Pk.
Material Polyester
Disposable Reusable
Lens Material Polyester
Frame Material Plastic
UV Protection None
Autoclavable Not Autoclavable