Learn More
Safety First Aid Evolution Plus Eyewash Kit with Mirror and 2 x 500 mL Eyewash Bottles
Product Code. 31759388
Change view
Click to view available options
Color:
Clear
Unit Size:
Each
Product Code. Color Unit Size
31759388 Clear Each
1 options
Product Code. 31759388 Supplier Safety First Aid Supplier No. E459M
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Evolution Plus Eyewash Kit with Mirror is a complete emergency eye care kit containing 2 x 500 mL sterile eyewash bottles and 2 sterile eye dressings.
  • The kit is supplied in a durable hinged case that includes bottle stabilisers to hold contents firmly in place.
  • It includes a quick release wall fixing bracket and kit identification label to provide a better storage and an easier identification.
  • Once on the bracket, the kit can be opened and worked with, providing a self-supporting workbench.
  • Evolution Plus Eyewash Kit assures you have adequate eye wash facilities at your place of work and is available for purchase, with or without mirror.
  • Evolution Plus Eye Wash with Mirror Contains:
    • 2x HypaClens Eye Wash Bottles, 500 mL
    • 2x HypaCover Sterile Eye Dressings
Specifications
Color Clear
Flow Rate 20 gpm (Shower), 8.0 gpm (Eye and Face Wash)
Material Plastic
Mounting Wall
Product Type Portable Eyewash and Shower Fixtures