Learn More
Safety First Aid Evolution Plus First Aid, Eye Wash and Burns Point, Large
Product Code. 31759391
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
Product Code. Sterility Unit Size
31759391 Sterile Each
1 options
Product Code. 31759391 Supplier Safety First Aid Supplier No. FAP37LG
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Evolution First Aid, Eye Wash and Burns Point (Large), British Standard Compliant is a complete first aid solution for large workplaces at risk of a wide range of injuries. It includes a first aid kit, an eyewash kit and a burns kit.
  • This First Aid Point is compliant with British Standard guidance for workplaces and provides the necessary content to treat a variety of injuries.
  • It comes fully stocked and includes brackets to ensure kits can be secured to the wall.
  • The kits are removable and portable, and the point has bracket labels to identify when a kit has been removed.
  • Photo-luminescent bracket headers allow for easy identification of the kit's location even in dark or smoky conditions.
  • Each kit features a shelf, ensuring contents remain in place when the kit is open on the wall.
  • The brackets allow each kit to be opened and used as a workbench, making the station ideal for all rapid response situations.
  • First Aid Kit Contents:
    • 1x First Aid Guidance Leaflet
    • 6x HypaCover First Aid Dressings, 12 x 12 cm
    • 4x HypaCover First Aid Dressings, 18 x 18 cm
    • 4x HypaBand Triangular Bandages
    • 4x HypaCover Eye Dressings
    • 100x HypaPlast Washproof Plasters
    • 40x HypaClean Sterile Wipes
    • 3x HypaPlast Microporous Tape, 2.5 cm
    • 12x HypaTouch Nitrile Gloves (Pair)
    • 4x HypaCover Finger Dressings
    • 2x HypaGuard Face Shields
    • 3x HypaGuard Foil Blankets
    • 2x Burn Dressings, 10 x 10 cm
    • 1x Clothing Cutters
    • 2x HypaBand Conforming Bandages, 7.5 cm
  • Eye Wash Kit Contents:
    • 2x HypaClens Eye Wash Bottles, 500 mL
    • 2x HypaCover Eye Dressings
  • Burns Kit Contents:
    • 1x First Aid Guidance Leaflet
    • 2x Burn Dressings, 10 x 10 cm
    • 1x Burn Dressing, 20 x 20 cm
    • 6x Burn Gel Sachets, 3.5 g
    • 12x HypaBand Safety Pins
    • 3x HypaBand Conforming Bandages
    • 2x HypaTouch Disposable Gloves (Pair)
    • 1x Clothing Cutters
Specifications
Product Type First Aid Kit
Material Plastic
Sterility Sterile