Learn More
Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometers
Shop All Thermo Scientific Products
Description
For routine, complex research and advanced applications, including current compliance, the Thermo Scientific™ Evolution™ One/One Plus UV-Vis Spectrophotometer series brings together reliable, versatile hardware, a broad range of accessories, with easy-to-use, easy-to-learn Thermo Scientific™ Insight™ Pro Software.
Webinar: Identification of protein aggregation through UV-Vis spectroscopy
For routine to research analysis, this instrument series offers real innovations that provide enhanced usability and performance - without added complexity. With straightforward software, cutting-edge instrumentation and a wide selection of accessories, the Evolution One and One Plus UV-Vis Spectrophometers work with you to perform experiments the way you want - for the results you need.
Optimized Engineering Delivers Ultimate Performance:- Double-beam geometry with fixed 1nm or variable spectral bandwidth options
- Application Focused Beam Geometry (AFBG) technology enables the best performance from microcells, solid samples, and fiber optic probes (Evolution One Plus Only)
- Xenon flash lamp requires no warm-up time allowing instant measurements
- Meet chemical, materials science, or biological sampling and measurement needs with a broad range of accessories
- Reduce training time: Optimized user interface with prompting software defines clear step-by-step workflow and is easy to teach and learn
- Prevent costly sampling and measurement errors: Create dedicated workflows for complex assay and transform multi-step workflows into one simplified, easy-to-follow method suitable for any level of user with Customized User Environment (CUE)
- Always current: Whether it is user inputs or application requirements, or new regulations, Insight Pro Software is continuously improving with quick upgrades to deliver greater capabilities and efficiencies for your lab’s needs
- Wide accessory selection with various temperature control options, sample changers, sippers and solid sampling devices complete your analysis
- Choice of Peltier Single Cell Holder or a Smart 8-Cell Peltier system for temperature control and sample monitoring from 0° to 100 °C
- Easy software integration gives you complete control of your measurements
Regulatory/Quality Control Environments:
To use the EvolutionOne/One Plus UV-Vis Spectrophotometers in a regulated or quality control environment, we offer a wide selection of compliance products and services to reduce implementation time and assure system performance.
- Evolution One or One Plus UV-Vis Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro US Ltr or INTL A4 Paper Version
- Qualification Filter Kit for UV-Vis
- Evolution One or One Plus Spectrometer
- INSIGHT Pro with Security Software
- Validator for Insight Pro US Ltr or INTL A4 Paper Version
- USP and EP UV Standards Set
- USP and EP Visible Standards Set
Specifications
Specifications
| Baseline Flatness | ±0.001 A, 200–800 nm, 1.0 nm SBW, smoothed |
| Detector Type | Dual Silicon Photodiodes |
| Dimensions (L x W x H) | 593 x 475 x 266 mm (23.3 x 18.7 x 10.6 in.) |
| Light Source | Xenon Flash |
| Product Line | Evolution One |
| Spectral Bandwidth | 1 nm |
| Type | Evolution One UV-Vis Spec |
| Wavelength Range | 190 to 1100 nm |
| Drift | <0.0005 A/hr., 500 nm, 1.0 nm SBW, 1 hour warmup |
| Electrical Requirements | 100/240 V, 50/60 Hz selected automatically, 150 W maximum |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
