missing translation for 'onlineSavingsMsg'
Learn More

Safety First Aid Evolution Workplace First Aid Kit, British Standard Compliant, Small

Product Code. 31759427
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
31759427 Sterile Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31759427 Supplier Safety First Aid Supplier No. K3031SM
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ensure your workplace is properly prepared for everyday first aid needs with the Evolution Workplace First Aid Kit (Small), fully compliant with British Standard BS8599-1 for small and lower-risk workplaces. Developed for UK businesses, this kit balances compact size with essential coverage for low-risk work places.

  • Why Choose This Kit?
    • All UK employers have a duty under the Health and Safety (First Aid) Regulations to provide adequate first aid provision.
    • This small Evolution kit meets minimum legal requirements for lower-risk environments and smaller staff numbers, offering peace of mind that your workplace is aligned with both legislation and best practice.
  • Key Benefits Features
    • Comprehensive, ready-to-use contents - includes bandages, plasters, wipes, dressings, scissors, tape, and more. No need to source individual items.
    • Durable, organised case - injection-moulded plastic box with a white cross and First Aid label for easy identification in an emergency.
    • Space-efficient design - measures 30 x 25 x 12 cm, easily stored on a shelf, in a cabinet, or in a small workplace cupboard.
    • Wall-mountable option - use the optional bracket for visible, easy access.
    • Versatile for many settings - ideal for offices, shops, salons, reception areas, service vans, and light industrial zones.
    • Maintains compliance - BS 8599-1 certification supports audits, inspections, and demonstrates commitment to workplace safety.
    • Easy to restock - refill packs and individual components are available to keep the kit up to date.
  • What BS 8599-1 Compliance Means
    • A kit compliant with BS 8599-1 meets nationally recognised standards for workplace first aid.
    • Contents and size are based on risk assessments, considering employee numbers and workplace hazards.
  • Recommended for:
    • 1 to 25 employees in low-risk workplaces
    • 1 to 5 employees in higher-risk settings
  • Tips for Use Maintenance
    • Regular checks: inspect monthly for expiry dates, used items, and proper storage.
    • Use original refills: maintain kit integrity and standardisation with matching refill packs.
    • Training integration: familiarise staff with the kit during first aid training sessions.
    • Strategic placement: position in visible, accessible areas (e.g., entrance, corridor, kitchen).
    • Records audits: keep a log of checks, restocks, and usage for compliance purposes.
  • The Evolution British Standard Compliant Workplace First Aid Kit in Green Case (small) Contains:
    • 1 x First Aid Guidance Leaflet
    • 2 x HypaCover First Aid Dressings, 12 x 12 cm
    • 2 x HypaCover First Aid Dressings, 18 x 18 cm
    • 2 x HypaBand Triangular Bandages
    • 2 x HypaCover Eye Dressings
    • 40 x HypaPlast Washproof Plasters
    • 20 x HypaClean Sterile Wipes
    • 1 x HypaPlast Microporous Tape, 2.5 cm x 5 m
    • 6 x HypaTouch Nitrile Gloves (pair)
    • 2 x HypaCover Finger Dressings
    • 1 x HypaGuard Face Shield
    • 1 x HypaGuard Foil Blanket
    • 1 x Burn Dressing, 10 x 10 cm
    • 1 x Clothing Cutters
    • 1 x HypaBand Conforming Bandage, 7.5 cm
  • Specifications
    • Dimensions: 30 x 25 x 12 cm
    • Case material: Injection-moulded plastic (robust and lightweight)
    • Compliance: BS 8599-1 for small workplaces
    • Optional wall mounting: Bracket sold separately
  • Ideal For
    • Small medium offices
    • Retail stores, salons, cafés, and customer-facing venues
    • Service vehicles, vans, or off-site employees
    • Low-risk workplaces (administration, meeting rooms, light workshops)
    • Health safety officers
    • Start-ups, remote locations, or break rooms

Specifications

Product Type First Aid Kit
Material Plastic
Sterility Sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.