Learn More
EVOS™ EVOS™ Condenser Slider, Green
Product Code. 15339651
Change view
Click to view available options
Quantity:
Each
Unit Size:
Each
Product Code. Quantity unitSize
15339651 Each Each
1 options
Product Code. 15339651 Supplier EVOS™ Supplier No. AMEPCFG
Only null left
Add to basket
Add to basket
Adds contrast when imaging
  • Used to improve illumination under certain conditions 
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Enables direct adjustment of white light LED color temperature
  • Intended for brightfield applications at all magnifications as daylight-correction filter
TRUSTED_SUSTAINABILITY
Specifications
Type Condenser Slider
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Diffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity Each