Learn More
EVOS™ EVOS™ Condenser Slider, Diffusion
Product Code. 15349651
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity unitSize
15349651 1 each Each
1 options
Product Code. 15349651 Supplier EVOS™ Supplier No. AMEPDFS1
Only null left
Add to basket
Add to basket
Diffuses light
  • Used to improve illumination under certain conditions
  • Due to variances in sample size, color and thickness, actual slider use may differ from suggestions
  • Intended for brightfield applications at 2x or 4x magnification for flat-field illumination
Diffusion condenser slider is included with EVOS™ XL Imaging System.
TRUSTED_SUSTAINABILITY
Specifications
Type Condenser Slider
Product Line EVOS
For Use With (Equipment) EVOS™ FL Color Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Includes Diffuser Slider, Pinhole Slider, Meniscus Sliders
Quantity 1 each