missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ Calibration Slide

Product Code. 17236573
Change view
Click to view available options
Quantity:
1 slide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17236573 1 slide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17236573 Supplier Invitrogen™ Supplier No. AMEP4720
Estimated Shipment: 24-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The EVOS Calibration Slide is used to calibrate three parameters of the system: field of view, parfocality, and parcentration.

The EVOS Calibration Slide is used to calibrate three parameters of the EVOS imaging system: field of view, parfocality, and parcentration. Calibrating the parfocality ensures that the sample stays in focus when the objective is changed, and calibrating the parcentration ensures that an object in the center of the field of view will stay in the center of the field no matter which objective is being used.

The calibration slide offers you these advantages:
• Easy, accurate, and precise calibration of your microscope
• Five X-Y squares divided by grids of: 1.00, 0.200, 0.100, 0.050, and 0.025 mm
• Seven different calibration circles: 4.000, 2.000, 1.000, 0.400, 0.200, 0.100, and 0.050 mm
• Precision-ground, optical-quality glass
• Protective storage case

The calibration procedure involves two distinct steps. First, the calibration crosshair on the screen is matched to the crosshairs on the calibration slide. Then, the diameters of the reference circles on the calibration slide are measured. Each step is performed using both the monochrome and the color camera. The total time required to calibrate an objective is about five minutes. More information can be found in the system’s manual. Note that the factory-installed objectives supplied with EVOS imaging systems have been pre-calibrated. There is no need to calibrate them again unless they are removed and need to be re-installed.

TRUSTED_SUSTAINABILITY

Specifications

Type EVOS Calibration
Product Line EVOS
Dimensions (L x W) 76 x 26 mm
For Use With (Equipment) EVOS™ FL Auto Imaging System, EVOS™ FL Auto 2 Imaging System, EVOS™ M5000 Imaging System, EVOS™ M7000 Imaging System
Quantity 1 slide
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.