Learn More
EVOS™ EVOS™ 40X Objective, fluorite, LWD, phase-contrast
Description
RMS-threaded objectives with 45mm parfocal distance
Specifications
Specifications
| Type | Objective Lens |
| Product Line | EVOS |
| For Use With (Equipment) | EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System |
| Lens Type | Plan Fluorite |
| Magnification | 40X |
| Numerical Aperture | 0.65 |
| Quantity | 1 each |