missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ M7000 Imaging System, High-Content Analysis Package with 3D

Product Code. 17816521
Change view
Click to view available options
Quantity:
1 package
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17816521 1 package Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17816521 Supplier Invitrogen™ Supplier No. AMF7000HCA3D
Power Your Renewable Energy Research
Use promo code 28029
View all promotions
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The EVOS M7000 Imaging System, High-Content Analysis Package with 3D, combines the fully automated, inverted, multi-channel fluorescence and transmitted light imaging system with Celleste 6.0 Image Analysis Software, a full-featured image analysis suite with protocols optimized to deliver the most relevant data for a range of popular live and fixed cell-based assays.

The EVOS M7000 Imaging System, High-Content Analysis Package with 3D, combines the fully automated, inverted, multi-channel fluorescence and transmitted light imaging system with Celleste 6.0 Image Analysis Software, a full-featured image analysis suite with protocols optimized to deliver the most relevant data for a range of popular live and fixed cell-based assays. The EVOS M7000 system was developed to meet today’s expectations for publication-quality images and data with just a few clicks using its powerful and time-saving acquisition tools with enhanced autofocus algorithms and automated routines for microwell plate assays, time-lapse live cell imaging, area scanning in time lapse mode, and Z-stacks in scan mode.

Celleste 6.0 software allows you to customize image analysis to your specific needs or take advantage of preconfigured analysis templates optimally designed to segment and classify images from a range of common cell-based assays. The union of the Celleste 6.0 software and EVOS microscope empowers you to easily transition from performing traditional microscopy to high-content analyses.

The EVOS M7000 High-Content Analysis packages offer:
• Highly configurable microscope—with a five-position objective turret and the ability to simultaneously acquire images in four fluorescence channels plus transmitted light with magnification from 1.25X to 100X, the optical system can be easily configured to meet your sample needs
• Publication-ready imagery in a few clicks—including autofocus, stitching, tiling, Z-stack capability, time-lapse imaging and single-click multichannel capture
• Intuitive acquisition interface—easily program the EVOS M7000 system to run well-plate scanning, time lapse experiments, and tile-stitch/montage area scans in Z-stack and/or time-lapse modes in your vessel of choice
• Streamlined image analysis—Celleste 6.0 icon-based wizard-drive workflow eliminates guesswork when using the preconfigured and optimized analysis templates for common applications to help derive the most relevant results from image sets

System and hardware specifications:
Optics: infinity‐corrected optical system; RMS‐threaded objectives with 45-mm parfocal distance
Imaging mode: fluorescence, brightfield, color brightfield, and phase contrast
Illumination: five-position chamber for four fluorescence light cubes plus brightfield; light cubes with integrated hard coated filter set and LED light source with >50,000 hour life; broad selection of standard and specialty light cubes
Imaging methods: single-color, multi-color, area scan with montage or tile-stitch, time lapse, Z-stacking, movie capture
Objective capacity: five‐position turret
Objectives (not included): wide selection of high‐quality LWD and coverslip‐corrected objectives
Condenser: 60-mm long working distance condenser, four-position turret with a clear aperture and three phase annuli
Stage: motorized X/Y scanning stage; travel range 120 mm x 80 mm with sub‐micron resolution, drop‐in inserts to receive vessel holders and lock-down holders to fix sample in place during long scans
Focus mechanism: automated focus mechanism with sub‐micron resolution
LCD display: 23-inch high‐resolution touch screen color monitor (also fully controllable via mouse); 3840 x 2160 pixel resolution
Cameras: high-sensitivity 3.2 MP (2048 x 1536) monochrome CMOS sensor with 3.45 μm pixel resolution

TRUSTED_SUSTAINABILITY

Specifications

Magnification Power 1.25X to 100X
Objective Types LWD, Fluorescence, Brightfield, Color Brightfield, Phase Contrast
For Use With (Application) Neurobiology, Immuno-Oncology, Live-Cell Imaging, 3 D Cell Imaging, High-Resolution Tile Scanning, Immunohistochemistry (IHC), High-Content Analysis (HCA)
Nosepiece 5 Position Turret
Shipping Condition Room Temperature
Lighting Type LED
Camera Type 3.2MP, Monochrome, CMOS
Display Type LCD
Model EVOS M7000
Product Line EVOS
Product Type Inverted Phase-contrast Fluorescent Microscope
Resolution 3840 x 2160 pixels
Quantity 1 package
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.