missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EVOS™ M7000 Imaging System

Product Code. 15918206
Change view
Click to view available options
Quantity:
1 system
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15918206 1 system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15918206 Supplier Invitrogen™ Supplier No. AMF7000
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. Se returpolicy

High-performance, fully automated, inverted, multi-channel fluorescence and transmitted light imaging system. Developed to meet today’s expectations for image quality, user interface interactions, data generation speed and flexibility, and end-user value with just a few clicks.

  • Outstanding image quality and versatility with a 5-position objective turret, 4-color LED fluorescence and transmitted light channels, and 3.2 MP CMOS color and B/W cameras
  • Exceptional usability with fully automated X/Y scanning stage, autofocus, and acquisition routines
  • High-speed image acquisition coupled with multi-position well scanning, and Z-stack and tile-stitch options lend power and flexibility to your data generation
  • Fully integrated time-lapse live cell imaging using the optional EVOS Onstage Incubator for precise control of temperature, gases for normoxic or hypoxic conditions, and humidity
  • Powerful image analysis capabilities for cell segmentation and quantification with the optional Celleste™ Image Analysis Software
  • Combines and integrates precision components with a unique modern design functionality that enables high-quality automated fluorescence imaging anywhere with unprecedented workflow efficiency. Full automation of the precision X/Y-stage movement; changing of the 4 LED fluorescent light cubes, 5-position objective turret, focus, and exposure; and switching of the dual camera make the EVOS M7000 system an exceptional platform for a variety of demanding imaging applications
  • Can program to run well-plate scanning, time lapse experiments, and tile-stitch/montage area scans in Z-stack and/or time-lapse modes in your vessel of choice
  • Compatible with all accessories for the predecessor EVOS FL Auto imaging system
  • Versatile and highly configurable with a 5-position objective turret and the ability to simultaneous acquire images in four fluorescence channels plus transmitted light
  • An extensive suite of tools for acquisition, visualization, and analysis
  • Smart LED illumination technology
  • Easy to use and reliable
  • *Note that three (3) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug: AMEP4644 EVOS Power Cord, Type A (North America) AMEP4645 EVOS Power Cord, Type G (United Kingdom) AMEP4646 EVOS Power Cord, Type C (Europe) AMEP4647 EVOS Power Cord, Type I (Australia) AMEP4648 EVOS Power Cord, Type L (Italy) AMEP4649 EVOS Power Cord, Type H (Israel) AMEP4708 EVOS Power Cord, Type D (India/South Africa) AMEP4782 Power Cord, Type I (China, CCC certified) AMEP4786 Power Cord Type J (Switzerland)

    TRUSTED_SUSTAINABILITY

    Tekniske data

    Type Cell Imaging System
    Product Line EVOS
    Camera 3.2 MP Color CMOS
    For Use With (Application) Neurobiology, Immuno-Oncology, Live-Cell Imaging, 3D Cell Imaging, High-Resolution Tile Scanning, Immunohistochemistry (IHC)
    Model M7000
    Light Source LED
    Magnification 1.25X to 100X
    Objectives 5 Position Turret
    Resolution 1920 x 1080 pixels
    Display LCD
    Content And Storage

    EVOS M7000 Imaging System

    • 27-inch 4K LCD monitor with 3840 x 2160 pixel resolution
    • External Dell computer with Intel Core i9 processor, 128 GB DDR5 RAM, 4 TB SSHD, and NVIDIA Quadro RTXA4000 with 16 GB discrete video graphics running Windows 11 LTSC

    EVOS M7000 Accessory Kit

    • EVOS M7000 Quick Start Guide
    • 16 GB USB 3.0 drive with user manual
    • Universal power supply, 24V, 5A
    • 3 North America (Type B) power cords*
    • USB 3.0 type A to B cable
    • Display port to display port cable, 6 ft
    • Cube access tool
    • EVOS calibration slide (AMEP4720)
    • Vessel holder with retention for two slides (AMEPVH021)
    • Vessel holder with retention for microwell plates (AMEPVH022). Also functions as adapter for vessel holders AMEPVH001 through AMEPVH018.
    • Block condenser slider (AMEP4688)
    • Diffuser condenser slider for brightfield applications (AMEPDFS1)
    • Slider for 4X objectives (AMEP4738)
    • EVOS light shield
    • Dust cover
    • Shipping restraint removal guide
    • Mouse pad
    • Tech Support card
    • Corded optical USB mouse
    • Keyboard

    * Note that three (3) country-specific power cord must be ordered separately in regions not using the Type B (North America) power plug:

    • AMEP4644 EVOS Power Cord, Type A (North America)
    • AMEP4645 EVOS Power Cord, Type G (United Kingdom)
    • AMEP4646 EVOS Power Cord, Type C (Europe)
    • AMEP4647 EVOS Power Cord, Type I (Australia)
    • AMEP4648 EVOS Power Cord, Type L (Italy)
    • AMEP4649 EVOS Power Cord, Type H (Israel)
    • AMEP4708 EVOS Power Cord, Type D (India/South Africa)
    • AMEP4782 Power Cord, Type I (China, CCC certified)
    • AMEP4786 Power Cord Type J (Switzerland)
    Quantity 1 system
    Shipping Condition Room Temperature
    Vis mere Vis mindre

    For Research Use Only. Not for use in diagnostic procedures.

    Produkttitel
    Vælg et problem

    Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.