Learn More
EVOS™ EVOS™ Vessel Holder, two 60-mm Petri dishes
Product Code. 11987315
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
Product Code. Quantity unitSize
11987315 1 each Each
1 options
Product Code. 11987315 Supplier EVOS™ Supplier No. AMEPVH003
Add to basket
Add to basket
For flasks, trays and petri dishes
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format two, 60 mm Petri dishes
Holds 2 60 mm Petri Dishes
Quantity 1 each