Learn More
Invitrogen™ EVOS™ Vessel Holder, one 100-mm Petri dish
Product Code. 11997315
Change view
Click to view available options
Quantity:
1 Holder
Unit Size:
Each
Product Code. Quantity Unit Size
11997315 1 Holder Each
1 options
Product Code. 11997315 Supplier Invitrogen™ Supplier No. AMEPVH004
Estimated Shipment: 24-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For flasks, trays and petri dishes
Broad range of holders helps maximize the flexibility of the EVOS and Micromaster™ inverted microscopes.
TRUSTED_SUSTAINABILITY
Specifications
Type Vessel Holder
Product Line EVOS
For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Format one 100-mm Petri dish
Holds 1 100 mm Petri Dish
Quantity 1 Holder