missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ex 26

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ex 26 is a potent and selective S1P1 receptor antagonist (IC50 = 0.93 nM). Exhibits > 3,000-fold selectivity for S1P1 over S1P2, S1P3, S1P4 and S1P5. Induces lymphocyte sequestration in peripheral lymph nodes and upregulates S1P1 receptor expression. Reduces disease severity in mice with EAE. Increases IFN-α production in stimulated dendritic cells in vivo.
Specifications
Specifications
| Chemical Name or Material | 1-[[[5'-[[1-(4-Chloro-3-methylphenyl)ethyl]amino]-2'-fluoro-3,5-dimethyl[1,1'-biphenyl]-4-yl]carbonyl]amino]-cyclopropanecarboxylic acid |
| CAS | 144678-63-7 |
| Quantity | 50 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C28H28ClFN2O3 |
| Formula Weight | 494.98 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction