missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ex 26
GREENER_CHOICE

Product Code. 18749947
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18749947

Brand: Tocris Bioscience™ 5833/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective S1P1 receptor antagonist

Ex 26 is a potent and selective S1P1 receptor antagonist (IC50 = 0.93 nM). Exhibits > 3,000-fold selectivity for S1P1 over S1P2, S1P3, S1P4 and S1P5. Induces lymphocyte sequestration in peripheral lymph nodes and upregulates S1P1 receptor expression. Reduces disease severity in mice with EAE. Increases IFN-α production in stimulated dendritic cells in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[[[5'-[[1-(4-Chloro-3-methylphenyl)ethyl]amino]-2'-fluoro-3,5-dimethyl[1,1'-biphenyl]-4-yl]carbonyl]amino]-cyclopropanecarboxylic acid
CAS 144678-63-7
Quantity 10 mg
Solubility Soluble to 100mM in DMSO and to 100mM in ethanol
Molecular Formula C28H28ClFN2O3
Formula Weight 494.98
Physical Form Solid
Color White
Product Type Bioactive Small Molecule
Purity >98%

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.