missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies EX527
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
- EX527 is a cell-permeable, selective inhibitor of mammalian sirtuin 1 (SIRT1; IC50 = 98 nM) over SIRT2 and SIRT3 and has no effect on other histone deacetylases (HDACs; Nayagam et al.) SIRT1 is a nicotinamide adenine dinucleotide (NAD)-dependent deacetylase with roles in energy metabolism and inflammation
- Studies have shown that EX527 inhibits sirtuins by forming a trimeric sirtuin complex with an NAD+-derived coproduct (Gertz M et al.)
- Increases the production of oligodendrocytes from differentiating neural stem cells and neural progenitor cells in vitro (Rafalski et al.)
- Restores the microvascular response during the hypoinflammatory phase in a mouse model of sepsis, and enhances the systemic innate immune response (Vachharajani et al.)
- Delays cyst growth in kidneys of PKD1 knockout mouse models (Zhou et al.)
Specifications
Specifications
| CAS | 49843-98-3 |
| Synonym | Selisistat |
| Quantity | 10 mg |
| Molecular Formula | C13H13ClN2O |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction