Learn More
Klever Excel Plus Metal Detectable
Product Code. 17739027
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 50
Product Code. Color Unit Size
17739027 Blue Pack of 50
1 options
Product Code. 17739027 Supplier Klever Supplier No. PLS40030MD
  • ONLINE EXCLUSIVE PRICE
    468.60 €
    404.50 € / Pack of 50
    Save 64.10 €
    13% Off
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Klever Excel Plus MD is a safety knife with a concealed blade and thus the highest level of safety. It protects people and material at the same time.
  • Due to the wider blade feeder, even thicker material can be cut easily and safely.
  • Due to the metal-detectable plastic, the safety knife is very well suited for use in the food, chemical or pharmaceutical industry.
  • The disposable product cuts up to double-wall corrugated cardboard, film, adhesive tape, strapping tape, etc.
  • With practical eyelet for lanyard fastening or hanging.
Specifications
Product Type Safety Knife
Disposable Single-use
Sterility Non-sterile
Color Blue