missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Exonuclease I (20 U/μL)

Product Code. 10354530 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
20,000 units
4,000 units
Unit Size:
20000 units
4000 units
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10354530 20,000 units 20000 units
10548930 4,000 units 4000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10354530 Supplier Thermo Scientific™ Supplier No. EN0582
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Degrade single-stranded DNA in a 3'→5' direction with Exonuclease I (ExoI), leaving 5'-terminal dinucleotides intact.

Exonuclease I (ExoI) degrades single-stranded DNA in a 3'≥5' direction. It releases deoxyribonucleoside 5'-monophosphates in a stepwise manner and leaves 5'-terminal dinucleotides intact.

It does not cleave DNA strands with terminal 3'-OH groups blocked by phosphoryl or acetyl groups.

Highlights

  • Active in PCR buffers

Applications

  • Primer removal from PCR mixtures:
    • prior to PCR product sequencing
    • li>for one-tube "megaprimer" PCR mutagenesis
  • Removal of single-stranded DNA containing a 3'-hydroxyl terminus from nucleic acid mixtures
  • Assay for the presence of single-stranded DNA with a 3'-hydroxyl terminus

Note

The enzyme is not suitable for removing 3'-overhangs of dsDNA.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 U/μL
Enzyme Exonuclease
Quantity 20,000 units
Product Type Exonuclease I
compliance-icons
Product Identifier
  • 10X Reaction Buffer for Exonuclease I
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: Potassium hydroxide

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.