Learn More
Descripción
Exonuclease I (ExoI) degrades single-stranded DNA in a 3'≥5' direction. It releases deoxyribonucleoside 5'-monophosphates in a stepwise manner and leaves 5'-terminal dinucleotides intact.
It does not cleave DNA strands with terminal 3'-OH groups blocked by phosphoryl or acetyl groups.
Highlights
- Active in PCR buffers
Applications
- Primer removal from PCR mixtures:
- prior to PCR product sequencing li>for one-tube "megaprimer" PCR mutagenesis
Note
The enzyme is not suitable for removing 3'-overhangs of dsDNA.
Especificaciones
Especificaciones
| Concentration | 20 U/μL |
| Enzyme | Exonuclease |
| Quantity | 4,000 units |
| Product Type | Exonuclease I |
Seguridad y manipulación
- Exonuclease I
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- MIXTURE LIST-Contain: Potassium hydroxide
For Research Use Only. Not for use in diagnostic procedures.
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.