missing translation for 'onlineSavingsMsg'
Learn More

Affymetrix™ ExoSAP-IT™ For PCR Product Clean-Up

Product Code. 15513687
Click to view available options
Quantity:
40 μL
Unit Size:
20 reactions
This item is not returnable. View return policy

Product Code. 15513687

Brand: Affymetrix™ 78250.40.UL

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.

  • Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
  • 30-minute protocol
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
For Use With (Application) Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs
No. of Reactions 20
Product Type Single-step enzymatic PCR Cleanup Kit
Quantity 40 μL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.