missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ ExoSAP-IT™ Express PCR Product Cleanup

Product Code. 15583677
Change view
Click to view available options
Quantity:
40 μL
Unit Size:
20 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15583677 40 μL 20 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15583677 Supplier Applied Biosystems™ Supplier No. 75001.40.UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ExoSAP-IT™ Express PCR Product Clean-up Reagent removes excess primers and unincorporated nucleotides from a PCR reaction. This enzymatic clean-up method offers precision, increased yield, and a faster turnaround time compared to alternative purification methods, such as spin columns or beads.

Key features of ExoSAP-IT™ Express reagent:

  • 5 minute protocol—Fastest turnaround time to results
  • One-tube, one-step PCR cleanup—Add ExoSAP-IT Express directly to PCR product
  • Novel enzyme technology—Irreversibly inactivated in just one minute at 80°C
  • Conserve PCR samples—100% recovery of PCR products, regardless of amplicon length
  • Eliminate spin columns or beads—Significantly decreases time and errors while increasing yield
  • Compatible—No interference in downstream applications
  • Scalable—Treat a wide range of volumes
  • Multiple formats—Single tube, 8 tube strip, and 96-well plate formats offer flexibility from manual pipetting to automated workflows
  • Go green—Generate less waste with a single-tube solution to PCR cleanup

Overview

It is necessary to remove excess primers and unincorporated nucleotides from the PCR reaction prior to sequencing.

An enzymatic cleanup method offers precision and speed over technologies such as spin columns or beads. ExoSAP-IT Express reagent consists of two hydrolytic enzymes, a modified Exonuclease I and Shrimp Alkaline Phosphatase (SAP), in a specially formulated buffer. It is used to remove excess primers and dNTPs from a PCR mixture with no interference in downstream applications. This unique, highly stable, one tube solution allows for greater confidence and superior sequencing results (Fig. 1).

Innovative solution

Our scientists engineered Exonuclease I to have increased heat lability, allowing for inactivation in only 1 minute (Fig. 2). ExoSAP-IT Express reagent is added directly to the PCR product for a 4 minute cleanup at 37°C, followed by a 1 minute inactivation at 80°C, which provides accurate and consistent results in just 5 minutes.

Fastest PCR cleanup method

This novel technology allows for a significant reduction in sample cleanup time with minimal steps, providing the simplest workflow (Fig. 3). ExoSAP-IT Express reagent offers rapid turnaround times and improved efficiency of resources while delivering the quality that scientists have become accustomed to when using ExoSAP-IT for PCR product cleanup.

Conserve PCR samples — simple one step, 100% recovery

The ExoSAP-IT Express enzymatic cleanup method is designed to minimize errors by reducing your protocol to a single pipette step, allowing for automated or manual processing from a single tube or microtiter well. ExoSAP-IT Express reagent outperforms the competition by ensuring 100% recovery of all amplicon sizes, both short and long (Table 1).

ExoSAP-IT Express PCR Product Cleanup is active in commonly used PCR buffers, so no buffer exchange is required. After treatment, ExoSAP-IT Express reagent is inactivated by heating at 80°C for 1 minute. The treated PCR products are then ready for subsequent analysis in applications that require DNA to be free of excess primers and nucleotides.

Specifications

Content And Storage Shipped on dry ice. Store at -20°C.
Format 8 Tube Strip, 96-well Plate, Tube
Formulation Liquid
For Use With (Application) PCR Purification
No. of Reactions 20 Reactions
Product Type Express PCR Product Clean-up Reagent
Quantity 40 μL
Test Time 5 min.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.