missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ ExoSAP-IT™ PCR Product Cleanup
Click to view available options
Quantity:
40 μL
Unit Size:
Each
Specifications
Specifications
| Description | ExoSAP-IT™ PCR Product Clean-up |
| Format | Spin Column |
| For Use With (Application) | PCR Purification |
| Quantity | 40 μL |
| Test Time | 15 min |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction