missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH EXPERT MOUNTING MEDIUM

Product Code. 30226293
Change view
Click to view available options
Volume (Metric):
1 L
4 x 250 mL
Unit Size:
1 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30226293 4 x 250 mL Pack of 4
30226292 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30226293 Supplier CELLPATH Supplier No. SEA160400A
  • ONLINE EXCLUSIVE PRICE
    151.00 €
    74.48 € / Pack of 4
    Save 76.52 €
    50% Off
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ExPert is a xylene-based medium, used for the mounting and subsequent preserving of slide specimens. Being a rapid-drying medium, it allows users to quickly screen slides within the laboratory.

  • ExPert possesses excellent long-term storage qualities, exhibiting no discolouration of the mounted specimen after years of archiving.
  • The refractive index is equivalent to the market leading product, being close to that of glass +20°C. Viscosity has been optimised to reduce the formation of air bubbles whilst increasing the medium's clarity.
  • Key Benefits:
  • Rapid drying
  • Suitable for manual and automated cover slipping
  • Refractive index equivalent to market leading equivalents
  • Viscosity is optimised to reduce air bubbles and increase clarity

Specifications

Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 4 x 250 mL
compliance-icons compliance-icons compliance-icons
Product Identifier
  • ExPert Mounting Medium
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use sand, carbon dioxideor powder extinguisher to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: Xylene, ethyl benzene, Toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.