missing translation for 'onlineSavingsMsg'
Learn More

CELLPATH EXPERT XTF MOUNTING MEDIUM

Product Code. 30226295
Change view
Click to view available options
Volume (Metric):
1 L
4 x 250 mL
Unit Size:
1 L
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30226295 4 x 250 mL Pack of 4
30226294 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30226295 Supplier CELLPATH Supplier No. SEA190400A
  • ONLINE EXCLUSIVE PRICE
    147.19 €
    68.19 € / Pack of 4
    Save 79.00 €
    53% Off
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

ExPert XTF is a xylene and toluene free mounting medium which is used to coverslip and permanently preserve thin stained tissue sections. Being xylene and toluene free (both of which can be carcinogenic), XTF provides enhanced safety to the end-user.

  • The refractive index is similar to ExPert being close to that of glass at +20°C.
  • Key Benefits:
  • Safe for end user as xylene and toluene free
  • Suitable for manual and automated cover slipping

Specifications

Product Type Mounting Medium
Reagent Type Mounting Solution
Volume (Metric) 4 x 250 mL
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • ExPert XTF
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P370+P378-In case of fire: Use sand, carbon dioxideor powder extinguisher to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: XTF Clearing Agent,Isopar E, Isobutyl methacrylate, 2-ethylhexyl methacrylate,1-dodecanethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.