missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ Inducible Expression System Kit

Product Code. 16386395
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16386395 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16386395 Supplier Gibco™ Supplier No. A39251
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Expi293 Inducible Expression System Kit is part of the Expi293 platform for rapid, high-yield transient protein expression from mammalian 293 cells

  • System kit is centered on engineered Expi293F cells that stably express the tetracycline repressor (TetR) protein, thus enabling tetracycline-regulated protein expression
  • Inducible cell line was established by the stable integration of the regulatory plasmid pcDNA6/TR, which encodes the TetR gene under the control of the human CMV promoter
  • This cell line is a powerful tool for tetracycline-inducible high-yield expression of recombinant proteins

Features:

  • Tetracycline-inducible protein expression with extremely low basal expression levels
  • Equivalent protein yields to parental Expi293F cells—up to 1 g/L
  • pcDNA5/TO expression vector for tetracycline-regulated expression of the gene of interest
  • Suspension, high-density culture of Expi293F Inducible cells in Expi293 Expression Medium
  • High-efficiency transient transfection using ExpiFectamine 293 reagent in combination with specialized transfection enhancers

Components included:

  • 1 vial of frozen Expi293F Inducible cells
  • 1 L of Expi293 Expression Medium
  • 1 ExpiFectamine 293 transfection kit sufficient to transfect 1 L of culture
  • 100 mL Opti-Plex Complexation Buffer
  • 1 pcDNA5/TO Mammalian Expression Vector kit
  • 500 mg tetracycline hydrochloride
  • pRABBIT IgG IRES-EmGFP positive control vector

All components of the Expi293 Inducible Expression System are also available for individual purchase

TRUSTED_SUSTAINABILITY

Specifications

Type Inducible Expression System Kit
Delivery Type Transfection
Quantity 1 Kit
Content And Storage • Expi293F Inducible Cells (1x107 cells/vial), store in vapor-phase liquid nitrogen
• 1 L Expi293 Expression Medium, store at 2–8°C (protect from light)
• ExpiFectamine 293 Transfection Kit for 1 L of culture, store at 2–8°C
• 100 mL Opti-Plex Complexation Buffer, store at 2–8°C (protect from light)
• 20 μg pcDNA5/TO Mammalian Expression Vector, store at 2–8°C or long term storage at -20°C.
• pRABBIT IgG IRES-EmGFP Positive Control Vector, store at 2–8°C or long term storage at -20°C.
• 500 mg Tetracycline Hydrochloride, store at -20°C

Shipping Condition Approved for shipment at Room Temperature or on Wet or Dry Ice
Starting Material DNA
Cell Line Expi293F™
Product Line Expi293
For Use With (Application) Protein Expression
Promoter CMV/TO

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.