missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Expi293™ PRO Expression Medium

Product Code. 31337532 Shop All Gibco Products
Change view
Click to view available options
Quantity:
1 L
10 L
20 L
6 x 1 L
Unit Size:
Pack of 6
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31337532 10 L -
31337527 1 L -
31337531 6 x 1 L -
18856517 6 x 1 L Pack of 6
31337530 20 L -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31337532 Supplier Gibco™ Supplier No. A4000135503
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Expi293 PRO Expression Medium supports rapid, high-yield protein production from HEK293 cells. Designed for scalable, high-density cultures with consistent expression across a variety of proteins, including complex and low-yield targets.

Gibco Expi293 PRO Expression Medium is a chemically defined, serum-free, protein-free medium for growth and transfection of suspension-adapted human embryonic kidney (HEK) 293 cells. It is specifically designed as a core component to support high density culture of Expi293 PRO cells in the Expi293 PRO Expression System for scalable transient protein expression.

  • Supports growth of suspension HEK 293 cultures to densities of over 15 x 10^6 cells/mL
  • Transfection-compatible serum-free suspension culture medium
  • Helps enable sustained, high level expression of high density transiently transfected cultures, achieving yields of up to 5 g/L of recombinant protein
  • Supports growth and transfection of HEK293 cells in culture formats of 1 mL to 10 L
  • Does not contain phenol red

Expi293 PRO Expression Medium is formulated with GlutaMAX-I reagent. It is ready to use, with no supplementation required. The chemically defined formulation results in high reproducibility and lot-to-lot reliability. Expi293 PRO Expression Medium contains no human or animal-origin components. The medium is not recommended for adherent cell cultures.

TRUSTED_SUSTAINABILITY

Specifications

Cell Line 293 (HEK)
Cell Type Embryonic Kidney Cells
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Product Type Expression Medium
Serum Level Serum-free
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives GlutaMAX
Without Additives No HEPES, No Phenol Red
Product Line Expi293 PRO
Quantity 10 L
Content And Storage Store in the dark at +2°C to +8°C
Culture Type Suspension Cell Culture
Show More Show Less

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.