missing translation for 'onlineSavingsMsg'
Learn More

Koettermann™ Exploris™ Tall Chemical Storage Cabinet

Product Code. 14895361
Change view
Click to view available options
Door Style:
Hinged
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Unit Size
14895361 Hinged Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14895361 Supplier Koettermann™ Supplier No. 37500008
 Restricted Item
Estimated Shipment: 10-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy

Storage of non-combustible, non-corrosive harmful substances.

  • Constructed of high-grade steel (1.0330), electrogalvanised, polyurethane coated
  • Soundproofed, double skin doors

Specifications

Dimensions (D x W x H) Interior 51.6 x 120 x 192 cm
Door Style Hinged
For Use With (Application) Storage of chemicals
No. of Doors 2
No. of Shelves 4
Shelf Type Removable
Color White
Product Type Storage Cabinet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.