missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, with premixed ROX

Product Code. 10124403
Change view
Click to view available options
No. of Reactions:
2500 Reactions
500 Reactions
Unit Size:
2500 reactions
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions Unit Size
10124403 2500 Reactions 2500 reactions
10226113 500 Reactions Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10124403 Supplier Invitrogen™ Supplier No. 1179001K
  • ONLINE EXCLUSIVE PRICE
    6870.00 €
    4122.00 € / 2500 reactions
    Save 2748.00 €
    40% Off
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EXPRESS One-Step SYBR GreenER kits are compatible with fast-cycling instruments for your SYBR-based qPCR experiments.

This kit comes with premixed ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with a separate tube of ROX reference dye are also available.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
  • SYBR GreenER dye
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed.

Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored.
Detection Method SYBR
Format Tube
PCR Method 1-step RT-qPCR
Polymerase Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Kit Type EXPRESS One-Step SYBR GreenER Kit
Product Line SYBR GreenER
Product Type Real Time PCR SYBR Master Mix
Quantity 2,500 reactions
Sufficient For 2500 Reactions
For Use With (Application) Gene Expression
No. of Reactions 2500 Reactions
Sample Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.