missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EXPRESS One-Step SYBR™ GreenER™ Kit, with premixed ROX
Description
This kit comes with premixed ROX reference dye. EXPRESS One-Step SYBR GreenER SuperMix kits with a separate tube of ROX reference dye are also available.
- Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
- The One-Step SuperScript™ III Reverse Transcriptase for one-step qRT-PCR
- SYBR GreenER dye
- UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination
Order Info
Guaranteed to be stable for 6 months when properly stored.
Specifications
Specifications
| Content And Storage | EXPRESS One-Step SYBR™ GreenER™ SuperMix Kits are composed of 2 modules: EXPRESS SYBR™ GreenER™ RT module (containing SuperScript™ III Reverse Transcriptase and RNAseOUT™) and EXPRESS SYBR™ GreenER™ qPCR SuperMix with ROX premixed. Store at -20°C upon receipt. Once thawed, the EXPRESS SYBR™ GreenER™ qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteed to be stable for 6 months when properly stored. |
| Detection Method | SYBR |
| Format | Tube |
| PCR Method | 1-step RT-qPCR |
| Polymerase | Taq DNA Polymerase |
| Reaction Speed | Fast or Standard |
| For Use With (Equipment) | 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode |
| Product Line | SYBR GreenER |
| Product Type | One-Step qRT-PCR Kit |
| Quantity | 500 reactions |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?