missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXPRESS qPCR Supermix, with premixed ROX

Product Code. 10174233
Change view
Click to view available options
No. of Reactions:
1000 Reactions
200 Reactions
Unit Size:
1000 reactions
200 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10174233 200 Reactions 200 reactions
10397333 1000 Reactions 1000 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174233 Supplier Invitrogen™ Supplier No. 11795200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

EXPRESS qPCR SuperMix kits are compatible with fast-cycling instruments for your probe-based qPCR assays.

  • Fast-activating, antibody-mediated Platinum™ Taq DNA Polymerase
  • UDG carryover protection - uracil-DNA-glycosylase (UDG)/dUTP to reduce carryover contamination

Order Info

Guaranteed minimum shelf life is 60 days.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage EXPRESS qPCR SuperMix kits are provided as a 2X qPCR SuperMix with ROX premixed or in a separate tube.

Store at -20°C upon receipt. Once thawed, the EXPRESS qPCR SuperMix can be stored for up to 1 month at +4·C. Product is guaranteedstable for 6 months when properly stored.

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
Detection Method Primer-probe
Format Tube
GC-Rich PCR Performance High
PCR Method qPCR
Polymerase Platinum Taq DNA Polymerase
Reaction Speed Fast or Standard
For Use With (Equipment) 7000 System, 7300 System, 7700 System, 7900HT Fast System, 7900HT System, Eppendorf™ MasterCycler™ RealPlex, StepOne™, Fast Mode, StepOne™, Standard Mode
Passive Reference Dye ROX (Pre-mixed)
Product Type Express qPCR Supermix
Quantity 200 reactions
Reverse Transcriptase SuperScript™ III
Sufficient For 200 Reactions
For Use With (Application) Gene Expression
No. of Reactions 200 Reactions
Sample Type DNA (Genomic), cDNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.