Learn More
Memmert™ Extension of DAkkS Factory Calibration for UF75plus
Extension of DAkkS factory calibration by an additional freely selectable temperature for Memmert Ovens.
Brand: Memmert™ UF75plusE39697
Additional Details : Weight : 0.01200kg
Description
- Extension of DAkkS calibration at Memmert factory by an additional freely selectable temperature value according to method C (DKD-R 5-7)
Specifications
UF75plus Oven | |
Factory Calibration Extension |