missing translation for 'onlineSavingsMsg'
Learn More

Tork Extra Hand Sanitizing Alcohol Gel​ Pump

Product Code. 30245605 Shop All Tork Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
30245605 Clear Pack of 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30245605 Supplier Tork Supplier No. 511205
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tork Extra Hand Sanitising Alcohol Gel effectively kills microorganisms to help prevent the spread of infections. The most effective gel in the Tork collection, with efficacy against bacteria, yeast and all viruses.

  • Effective against a wide range of pathogens - complies with EN 1500, EN 12791, and EN 14476.
  • Dermatologically tested. Suitable for frequent use
  • Time saving for cleaning staff: certified effortless refilling in less than 10 seconds
  • Ensure good hygiene: factory-sealed bottle with a new pump for each refill reduced
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hand Sanitizer
Color Clear
Form Gel
Active Ingredients Alcohol
Certifications/Compliance EN1500, EN12791, EN14476
Packaging Pump Bottle
Quantity 8/Pk.
Size 500 mL
compliance-icons compliance-icons
Product Identifier
  • Tork Extra Hand Sanitizing Alcohol Gel
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.