Learn More
Description
This package includes supplemental reagents for use with the MagMAX FFPE DNA/RNA Ultra Kit (Cat. No. A31881). These additional reagents consist of protease digestion buffer, binding solution, and DNA wash buffer and are specifically for use when processing >40 μm sections. See the User Guide for details.
Specifications
Specifications
| Content And Storage | • 10 mL protease digestion buffer, store at room temperature • 38.5 mL binding solution, store at room temperature • 38.5 mL DNA wash buffer, store (at room temperature) |
| Sample Type | FFPE, Fixed Samples |
| For Use With (Application) | Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | High-throughput Compatible, Manual Protocols, Automated Protocols |
| Product Line | MagMAX |
| Product Type | FFPE DNA/RNA Ultra Reagent |
| Quantity | 10 mL kit |
| Shipping Condition | Approved for shipment at Room Temperature. |
| Target | Genomic DNA, Total RNA |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.