Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Product Code. 10223911
Change view
Click to view available options
Standard Taper Joint Bottom:
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Bottom unitSize
10223911 NS 29/32 Each
10283951 NS 34/35 Each
10375971 NS 45/40 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10223911 Supplier Lenz Laborglasintrumente Supplier No. 05380025
  • ONLINE EXCLUSIVE PRICE
    741.00 €
    489.46 € / Pack of 1
    Save 251.54 €
    33% Off
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor head with vacuum jacket
  • Made of DURAN™ tubing
  • Twisselmann condenser (improved) with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 30 mL
Material Duran Glass
Standard Taper Joint Bottom NS 29/32