Learn More
Lenz Laborglasintrumente Extractors Acc. to Twisselmann
Product Code. 10375971
Change view
Click to view available options
Standard Taper Joint Bottom:
NS 29/32
NS 34/35
NS 45/40
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Bottom Unit Size
10375971 NS 45/40 Each
10223911 NS 29/32 Each
10283951 NS 34/35 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10375971 Supplier Lenz Laborglasintrumente Supplier No. 05380037
  • ONLINE EXCLUSIVE PRICE
    856.00 €
    603.52 € / Pack of 1
    Save 252.48 €
    29% Off
Estimated Shipment: 03-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Extractor head with vacuum jacket
  • Made of DURAN™ tubing
  • Twisselmann condenser (improved) with thread GL 14
  • Round bottom flask
Specifications
Capacity (Metric) 100 mL
Material Duran Glass
Standard Taper Joint Bottom NS 45/40