missing translation for 'onlineSavingsMsg'
Learn More

Ekastu Safety™ ECO Eye Wash Bottle with Funnel

Product Code. 10477073
Change view
Click to view available options
Capacity (Metric):
600 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
10477073 600 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10477073 Supplier Ekastu Safety™ Supplier No. 177111
Estimated Shipment: 18-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Made for immediate rinsing of the eyes after accidental splashing from acids, bases and other hazardous chemicals. EXASTU Safety™ ECO Eye Wash Bottle with Funnel is filled with water and can be used in both upright and horizontal positions.

  • DIN EN 15154-4
  • Filled (600mL)
  • Can be stored -maintanance free-
  • For immediate rinsing of the eyes after corrosion by acids, basis etc.
  • PE bottle with screwed-on eye funnel without tilting valve
  • Can be used in upright and horizontal position
  • Leak-proof
  • Economical construction
  • Without retail packaging
  • With transportation lock band
  • Our tip for storage: Eye Wash Station (EPS) empty (Cat no. 277 955)

Specifications

Capacity (Metric) 600 mL
For Use With (Application) Made for Immediate Rinsing of the Eyes after Accidental Splashing from Acids, Bases and other Hazardous Chemicals
Includes Leakproof Screw Cap with Eye Funnel
Material Polyethylene (Bottle)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.